missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole, 99%

Product Code. 11459143
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459143 25 g 25g
11439143 1 g 1g
11449143 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459143 Supplier Thermo Scientific Alfa Aesar Supplier No. A11665.14
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 271-44-3 | C7H6N2 | 118.139 g/mol

1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

Solubility
Soluble in hot water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-44-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00005691
InChI Key BAXOFTOLAUCFNW-UHFFFAOYSA-N
Synonym indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
PubChem CID 9221
ChEBI CHEBI:36669
IUPAC Name 1H-indazole
SMILES C1=CC=C2C(=C1)C=NN2

Specifications

Melting Point 146°C to 149°C
Boiling Point 270°C
Quantity 25 g
Beilstein 109676
Merck Index 14,4936
Solubility Information Soluble in hot water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 1H-Indazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.