missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10031694
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10031694 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10031694 Supplier Thermo Scientific Maybridge Supplier No. CC12701DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 771-50-6
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7NO2
Quantity 1g
Synonym indole-3-carboxylic acid, 3-indoleformic acid, 3-carboxyindole, indole-3-carboxylicacid, 3-indolecarboxylic acid, indole-3-carboxylate, 3-indolylcarboxylic acid, beta-indolylcarboxylic acid, indole-3 carboxylic acid, ico
InChI Key KMAKOBLIOCQGJP-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)O
IUPAC Name 1H-indole-3-carboxylic acid
Molecular Weight (g/mol) 161.16
PubChem CID 69867
ChEBI CHEBI:24809
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1H-Indole-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.