missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-7-carbohydrazide, 97%, Thermo Scientific™

Product Code. 10384342
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10384342 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10384342 Supplier Thermo Scientific Maybridge Supplier No. SB02108CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 321309-24-4
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H9N3O
MDL Number MFCD00572868
Quantity 250 mg
Synonym 1h-indole-7-carboxylic acid hydrazide, 7-hydrazinocarbonyl-1h-indole, 1h-indole-7-carboxylic acid, hydrazide, indole-7-carbohydrazide, 1h-indole-7-carboxylicacid, hydrazide
InChI Key NCFMBDUFIJHHAW-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C1)C(=O)NN)NC=C2
IUPAC Name 1H-indole-7-carbohydrazide
Molecular Weight (g/mol) 175.191
PubChem CID 712457
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 1H-indole-7-carbohydrazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.