missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-7-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10532272
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10532272 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10532272 Supplier Thermo Scientific Maybridge Supplier No. BTB12826CB
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1670-83-3
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C9H7NO2
MDL Number MFCD00210442
Quantity 250 mg
Synonym indole-7-carboxylic acid, 7-indolecarboxylic acid, indole-7-carboxylicacid, zlchem 444, pubchem2475, 7-carboxyindole, acmc-209dvl, indole-7-carboxlic acid, indole 7-carboxylic acid, 1h-indole-7-carboxylicacid
InChI Key IPDOBVFESNNYEE-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C1)C(=O)O)NC=C2
IUPAC Name 1H-indole-7-carboxylic acid
Molecular Weight (g/mol) 161.16
PubChem CID 74281
Formula Weight 161.16
Percent Purity 90%
Chemical Name or Material 1H-Indole-7-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.