missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-1-carboxamidine monohydrochloride, 99%

Product Code. 10570362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10570362 10 g Glass bottle 10g
10603773 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10570362 Supplier Thermo Scientific Acros Supplier No. 368250100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4023-02-3 | C4H6N4·HCl | 146.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-02-3
Molecular Formula C4H6N4·HCl
Molecular Weight (g/mol) 146.58
MDL Number MFCD00210087
InChI Key UCQFSGCWHRTMGG-UHFFFAOYSA-N
Synonym 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl
PubChem CID 2734672

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 167°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 146.58
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1H-Pyrazole-1-carboxamidine monohydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1H-Pyrazole-1-carboxamidine monohydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.