missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole, 98%

Product Code. 11439080
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439080 25 g 25g
11449080 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439080 Supplier Thermo Scientific Alfa Aesar Supplier No. A14186.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

1H-Pyrazole, 98%, C3H4N2, CAS Number-288-13-1, 1-h-pyrazole, 1h-pyrazol, pyrazole, unii-3qd5kjz7zj, hpz, pyrazol, diazole, pyrazol#1, 1,2-diazole, 3qd5kjz7zj, 100g, 103775, 186 deg.C to 188 deg.C, CHEBI:17241, 206-017-1, 68.08, H302-H315-H319-H335 | CAS: 288-13-1 | C3H4N2 | 68.079 g/mol

1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-13-1
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005234
InChI Key WTKZEGDFNFYCGP-UHFFFAOYSA-N
Synonym pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
PubChem CID 1048
ChEBI CHEBI:17241
IUPAC Name 1H-pyrazole
SMILES C1=CNN=C1

Specifications

Melting Point 66°C to 70°C
Boiling Point 186°C to 188°C
Odor Unpleasant
Quantity 25 g
Beilstein 103775
Sensitivity Hygroscopic
Merck Index 14,7960
Solubility Information Soluble in water.
Formula Weight 68.08
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.