missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole, TRC

Product Code. 30424473
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424473 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424473 Supplier Toronto Research Chemicals Supplier No. TRCP8421955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrazole
CAS 288-13-1
Molecular Formula C3 H4 N2
Quantity 5 g
Synonym Pyrazole, 1H-Pyrazole, 1,2-Diazole, 2-Azapyrrole, NSC 45410
InChI Formula InChI=1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)
SMILES c1cn[nH]c1
IUPAC Name 1H-pyrazole
Molecular Weight (g/mol) 68.08
Formula Weight 68.0374
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.