missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole, TRC

Product Code. 30417807
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417807 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417807 Supplier Toronto Research Chemicals Supplier No. TRCP84219550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Pyrazole
CAS 288-13-1
Molecular Formula C3 H4 N2
Quantity 50 g
Synonym Pyrazole, 1H-Pyrazole, 1,2-Diazole, 2-Azapyrrole, NSC 45410
InChI Formula InChI=1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)
SMILES c1cn[nH]c1
IUPAC Name 1H-pyrazole
Molecular Weight (g/mol) 68.08
Formula Weight 68.0374
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.