missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrrole-2-carbohydrazide, 97%, Thermo Scientific™

Product Code. 10384582
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10384582 250 mg Amber Glass Bottle 250mg
10243182 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10384582 Supplier Thermo Scientific Maybridge Supplier No. RF02986CB
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50269-95-9
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.131
MDL Number MFCD00141885
InChI Key FQIAWPZQJCAPSW-UHFFFAOYSA-N
Synonym pyrrolecarbohydrazide, 2-pyrrolecarbohydrazide, pyrrole-2-carbohydrazide, pyrrole-5-carboxhydrazide, 2-pyrrolecarboxylic acid hydrazide, 2-pyrrolecarboxylic acid, hydrazide, pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylicacid, hydrazide, 1h-pyrrole-2-carboxylic acid, hydrazide 9ci
PubChem CID 281610
IUPAC Name 1H-pyrrole-2-carbohydrazide
SMILES C1=CNC(=C1)C(=O)NN

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 1H-pyrrole-2-carbohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.