missing translation for 'onlineSavingsMsg'
Learn More

1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt, 97%, Thermo Scientific™

Product Code. 11405457
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405457 25 g 25g
11495447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405457 Supplier Thermo Scientific Alfa Aesar Supplier No. B23461.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96107-94-7
Molecular Formula C4H6N4NaO2
Molecular Weight (g/mol) 165.108
MDL Number MFCD02683105
InChI Key WDUYZCSZENDSPG-UHFFFAOYSA-N
Synonym sodium ethyl 1h-1,2,3,4-tetrazole-5-carboxylate
PubChem CID 67498453
IUPAC Name ethyl 2H-tetrazole-5-carboxylate;sodium
SMILES CCOC(=O)C1=NNN=N1.[Na]

Specifications

Melting Point ∼240°C (decomposition)
Assay Percent Range 0.99
Quantity 25 g
UN Number UN1325
Formula Weight 164.11
Percent Purity 97%
Chemical Name or Material 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt
compliance-icons
Product Identifier
  • 1H-Tetrazole-5-carboxylic acid ethyl ester,sodium salt
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.