missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Boric Acid
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
Description
For use in preparing electrophoresis buffers.
For use in preparing electrophoresis buffers.
Specifications
Specifications
| Chemical Name or Material | Boric Acid |
| CAS | 10043-35-3 |
| Quantity | 1 kg |
| Molecular Formula | H3BO3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction