missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments

Product Code. 15225753 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Hydroxylamine
Triethylammonium Bicarbonate (TEAB)
Unit Size:
50mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15225753 Hydroxylamine 5mL
15215753 Triethylammonium Bicarbonate (TEAB) 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15225753 Supplier Thermo Scientific™ Supplier No. 90115
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Common TMT buffers such as 1M Triethylammonium bicarbonate (TEAB) and 50% Hydroxylamine for isobaric mass tag labeling experiments.

Pre-formulated TEAB and 50% hydroxylamine designed for use in TMT labeling and quenching of peptides.

Triethylammonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hydroxylamine
Product Line TMT
Content And Storage Store protected from direct sunlight in a dry, cool, and well-ventilated area.
Starting Material Cell Lysate
Workflow Step Protein Labeling
Quantity 5 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydroxylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.