missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Benzenetricarboxylic Acid 2,4-Bis(2-ethylhexyl) Ester(Mixture of Diastereomers), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Benzenetricarboxylic acid, 2,4-bis(2-ethylhexyl) ester |
| CAS | 63468-11-1 |
| Molecular Formula | C25H38O6 |
| Quantity | 1 mg |
| Synonym | 1,2,4-Benzenetricarboxylic Acid 2,4-Bis(2-ethylhexyl) Ester(Mixture of Diastereomers) |
| InChI Formula | InChI=1S/C25H38O6/c1-5-9-11-18(7-3)16-30-24(28)20-13-14-21(23(26)27)22(15-20)25(29)31-17-19(8-4)12-10-6-2/h13-15,18-19H,5-12,16-17H2,1-4H3,(H,26,27) |
| SMILES | O=C(O)C1=C(C(OCC(CC)CCCC)=O)C=C(C(OCC(CC)CCCC)=O)C=C1 |
| IUPAC Name | 2,4-bis(((2-ethylhexyl)oxy)carbonyl)benzoic acid |
| Molecular Weight (g/mol) | 434.57 |
| Formula Weight | 434.27 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction