missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzotriazole-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Benzotriazole-d4 |
| CAS | 1185072-03-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 D4 H N3 |
| Quantity | 1 mg |
| Synonym | 1H-Benzotriazole-4,5,6,7-d4, 1H-Benzotriazole D4 (Phenyl D4) |
| InChI Formula | InChI=1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c([2H])c2[nH]nnc2c1[2H] |
| IUPAC Name | 4,5,6,7-tetradeuterio-1H-benzotriazole |
| Molecular Weight (g/mol) | 123.149 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction