missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(butoxyethyl) Phosphate-d8, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Bis(butoxyethyl) Phosphate-d8 |
| CAS | 2305665-16-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H19 D8 O6 P |
| Quantity | 1 mg |
| Synonym | 2-Butoxy-ethanol Hydrogen Phosphate-d8, Bis(2-butoxyethyl) Phosphate-d8, 2-Butoxy-ethanol 1,1'-(Hydrogen Phosphate)-d8 |
| InChI Formula | InChI=1S/C12H27O6P/c1-3-5-7-15-9-11-17-19(13,14)18-12-10-16-8-6-4-2/h3-12H2,1-2H3,(H,13,14)/i9D2,10D2,11D2,12D2 |
| SMILES | O=P(OC([2H])([2H])C([2H])([2H])OCCCC)(O)OC([2H])([2H])C([2H])([2H])OCCCC |
| IUPAC Name | bis(2-butoxy-1,1,2,2-tetradeuterioethyl) hydrogen phosphate |
| Molecular Weight (g/mol) | 306.36 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction