missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol Z-13C12, TRC

Product Code. 17972985
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972985 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972985 Supplier Toronto Research Chemicals Supplier No. TRCB5196521MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C613C12H20O2
Molecular Weight (g/mol) 280.26
Synonym 1,1-Bis(4-hydroxyphenyl)cyclohexane-13C12, 4,4'-Cyclohexylidenebis-phenol-13C12, 4,4'-Cyclohexylidenedi-phenol-13C12, 1,1-Bis(p-Hydroxyphenyl)cyclohexane-13C12, 4,4'-Cyclohexylidenebisphenol-13C12, 4,4'-Cyclohexylidenediphenol-13C12, Antigene W-13C12, Bis-Z-13C12, Dian C-13C12, NSC 29881-13C12, NSC 50761-13C12
IUPAC Name 4,4'-(cyclohexane-1,1-diyl)bis(phenol-13C6)
SMILES O[13C]1=[13C]([H])[13C]([H])=[13C](C2(CCCCC2)[13C]3=[13C]([H])[13C]([H])=[13C](O)[13C]([H])=[13C]3[H])[13C]([H])=[13C]1[H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H20O2/c19-16-8-4-14(5-9-16)18(12-2-1-3-13-18)15-6-10-17(20)11-7-15/h4-11,19-20H,1-3,12-13H2/i4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,14+1,15+1,16+1,17+1
Formula Weight 280.19
Percent Purity >95%
Chemical Name or Material Bisphenol Z-13C12
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.