missing translation for 'onlineSavingsMsg'
Learn More

CA-074 methyl ester, MedChemExpress

Product Code. 16420639
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16420639

Brand: Medchem Express HY100350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.

CA-074 methyl ester is a specific inhibitor of Cathepsin B, which has potent bioactivities such as neuroprotective, anti-cancer, and anti-inflamatory effects.

Specifications

Chemical Name or Material CA-074 methyl ester
CAS 147859-80-1
Color White
Purity Grade Notes Research
Molecular Formula C19H31N3O6
Quantity 1 mg
Synonym CA-074Me
Solubility Information DMSO : 100 mg/mL (251.59 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C([C@@H]1[C@@H](C(NCCC)=O)O1)N[C@]([C@@H](C)CC)([H])C(N(CCC2)[C@@H]2C(OC)=O)=O
Molecular Weight (g/mol) 397.47
Formula Weight 397.47
Percent Purity 98.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.