missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine-2,3,3,4,4,5,5-d7 HCl, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | L-Arginine d7 hydrochloride |
| CAS | 204244-77-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 D7 H7 N4 O2 . H Cl |
| Quantity | 1 mg |
| Synonym | L-Arginine-2,3,3,4,4,5,5-d7, hydrochloride (1:1), L-Arginine-2,3,3,4,4,5,5-d7, monohydrochloride (9CI), L-Arginine-2,3,3,4,4,5,5-d7 hydrochloride (1:1), L-Arginine-d7 hydrochloride, L-Arginine-2,3,3,4,4,5,5-D7 hydrochloride, (2S)-2-Amino-5-[(diaminomethylidene)amino]pentanoic-2,3,3,4,4,5,5-d7 acid hydrochloride, (2S)-2-Amino-5-carbamimidamidopentanoic-d7 acid hydrochloride, (S)-2-Amino-5-guanidinopentanoic-2,3,3,4,4,5,5-d7 acid hydrochloride, (S)-Arginine-d7 hydrochloride |
| InChI Formula | InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i1D2,2D2,3D2,4D; |
| SMILES | Cl.[2H]C([2H])(NC(=N)N)C([2H])([2H])C([2H])([2H])[C@]([2H])(N)C(=O)O |
| IUPAC Name | (2S)-2-amino-2,3,3,4,4,5,5-heptadeuterio-5-guanidino-pentanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 217.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction