missing translation for 'onlineSavingsMsg'
Learn More

WAY-204688, MedChemExpress

Product Code. 16471819
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16471819

Brand: Medchem Express HY19498

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress WAY-204688 is an estrogen receptor (ER-α) selective, orally active inhibitor of NF-κB transcriptional activity with an IC50 of 122 ± 30 nM for NF-κB-luciferase (NF-κB-luc) in HAECT-1 cells.

WAY-204688 is an estrogen receptor (ER-α) selective, orally active inhibitor of NF-κB transcriptional activity with an IC50 of 122 ± 30 nM for NF-κB-luciferase (NF-κB-luc) in HAECT-1 cells.

Specifications

Chemical Name or Material WAY-204688
CAS 796854-35-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C34H31F3N2O2
Quantity 1 mg
Synonym SIM-688
Solubility Information DMSO : 100 mg/mL (179.66 mM; Need ultrasonic)
SMILES COC1=C([C@H](C2=CC=CC3=C2C=CC=C3)[C@@](C)(C#N)C(N4CCC(C5=CC(C(F)(F)F)=CC=C5)CC4)=O)C=CC=C1
Molecular Weight (g/mol) 556.62
Formula Weight 556.62
Percent Purity 96.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.