missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human J Chain Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69762
This item is not returnable.
View return policy
Description
J Chain Polyclonal antibody specifically detects J Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Dot Blot, Immunocytochemistry
Specifications
| J Chain | |
| Polyclonal | |
| HRP | |
| IgJ chain;Immunoglobulin J chain;immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides;J chain;Joining chain of multimeric IgA and IgM | |
| Human J chain is a polypeptide folded within the structure of the polymeric immunoglobulin, resulting in a very limited exposure of J chain antigenic determinants. The antiserum is raised against the isolated and purified J chain. J chains isolated from polymeric IgA and IgM are identical by criteria of composition, peptide maps and antigenicity; Human J chain is distinct from all other chain components of polymeric IgA and IgM. It has a unique primary structure as shown by sequence analyses. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Human | |
| Lyophilized |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Dot Blot, Immunocytochemistry | |
| LYOPH | |
| Lyophilized from PBS,(pH 7.2) | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by adding 1 mL sterile distilled water, spin down to remove insoluble particles. | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction