missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%

Product Code. 10131192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10131192 1 g Glass bottle 1g
10167503 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10131192 Supplier Thermo Scientific Acros Supplier No. 364640010
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 20439-47-8 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20439-47-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062985,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES NC1CCCCC1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 40.0°C to 45.0°C
Color White to Yellow
Boiling Point 110.0°C to 114.0°C (40.0 Mbar)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NH2)2
Quantity 1 g
Beilstein 13, III, 6
Specific Rotation Condition − 24.50 (26.50°C c=5 1 N HCl 20°)
Specific Rotation − 24.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 114.19
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material (1R, 2R)-(-)-1, 2-Diaminocyclohexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (1R,2R)-(-)-1,2-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.