missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate, 98%

Product Code. 15446648
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446648 2 g 2g
15405536 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446648 Supplier Thermo Scientific Alfa Aesar Supplier No. H60172.04
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39961-95-0 | C10H20N2O6 | 264.278 g/mol

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39961-95-0
Molecular Formula C10H20N2O6
Molecular Weight (g/mol) 264.278
MDL Number MFCD00191979
InChI Key GDOTUTAQOJUZOF-ZXZVGZDWSA-N
Synonym 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n
PubChem CID 11448443
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 273°C (decomposition)
Quantity 2 g
Optical Rotation +12° (c=4 in Water)
Formula Weight 264.28
Percent Purity 99%
Chemical Name or Material (1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.