missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%

Product Code. 15439785
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439785 250 mg 250mg
15429147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439785 Supplier Thermo Scientific Alfa Aesar Supplier No. H52418.MD
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26685-83-6 | C7H13NO2 | 143.186 g/mol

(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26685-83-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD08276263
InChI Key USQHEVWOPJDAAX-PHDIDXHHSA-N
Synonym 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid #
PubChem CID 2724640
IUPAC Name (1R,2R)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Solubility Information Soluble in water.
Quantity 250 mg
Formula Weight 143.19
Optical Rotation −77° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material (1R,2R)-2-Aminocyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.