missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%

Product Code. 15435466
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435466 250 mg 250mg
15425466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435466 Supplier Thermo Scientific Alfa Aesar Supplier No. H60084.MD
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 452304-59-5 | C18H22NP | 283.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452304-59-5
Molecular Formula C18H22NP
Molecular Weight (g/mol) 283.36
MDL Number MFCD17013996
InChI Key ZATLZEHZPXYMFE-YTXKPMTANA-N
Synonym 1r,2r-2-diphenylphosphino cyclohexanamine, 1r,2r-2-diphenylphosphanyl cyclohexan-1-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 1r,2r-2-diphenylphosphino cyclohexylamine, 1r-2alpha-diphenylphosphino cyclohexane-1beta-amine, 1r,2r---2-diphenylphosphino cyclohexylamine, 97+%, 1r,2r-2-diphenylphosphino-1-aminocyclohexane, 1r,2r-2-diphenylphosphino-1-aminocyclohexane,min
PubChem CID 11166387
SMILES N[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 43°C to 48°C
Flash Point >110°C (230°F)
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Optical Rotation −79° (c=0.5 in Chloroform)
IUPAC Name (1R,2R)-2-diphenylphosphanylcyclohexan-1-amine
Formula Weight 283.35
Percent Purity ≥97%
Chemical Name or Material (1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.