missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, 90%, Thermo Scientific™

Product Code. 10681125 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10681125 1 g Glass bottle 1g
10127153 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10681125 Supplier Thermo Scientific Acros Supplier No. 379350010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55739-58-7
Molecular Formula C28H28O2P2
Molecular Weight (g/mol) 458.48
MDL Number MFCD05863546
InChI Key QKZWXPLBVCKXNQ-UHFFFAOYNA-N
Synonym r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
PubChem CID 10884975
IUPAC Name (R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
SMILES COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 102.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 3333
Specific Rotation -80° to -85° (20°C, 589nm) (C=1, CHCl3)
Formula Weight 458.48
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.