missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15409515
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15409515 1 g 1g
15419515 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15409515

Brand: Thermo Scientific Alfa Aesar H52022.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161660-94-2
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD01320858,MFCD01320857
InChI Key RNJQBGXOSAQQDG-SFYZADRCSA-M
Synonym 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
PubChem CID 1512526
SMILES CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

Specifications

IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
Quantity 1 g
Formula Weight 229.28
Percent Purity 95%
Chemical Name or Material (1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.