missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-Camphoric Acid, TRC

Product Code. 17934315
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934315 1 g 1g
17901225 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934315 Supplier Toronto Research Chemicals Supplier No. TRCC1659051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-83-4
Formula Weight 200.1049
InChI Formula InChI=1S/C10H16O4/c1-9(2)6(7(11)12)4-5-10(9,3)8(13)14/h6H,4-5H2,1-3H3,(H,11,12)(H,13,14)/t6-,10+/m1/s1
IUPAC Name (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
SMILES CC1(C)[C@H](CC[C@@]1(C)C(=O)O)C(=O)O
Synonym (1R,3S)-1,2,2-Trimethyl-1,3-cyclopentanedicarboxylic Acid, (1R-cis)-1,2,2-Trimethyl-1,3-cyclopentanedicarboxylic Acid, (1R,3S)-(+)-Camphoric Acid, (+)-(1R,3S)-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, (+)-Camphoric Acid, (1R)-cis-Camphoric Acid, D-(+)-Camphoric Acid, D-Camphoric aAid, Dextrocamphoric Acid, d-(+)-Camphoric Acid, d-Camphoric Acid;
Chemical Name or Material (1R,3S)-Camphoric Acid

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.