missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

Product Code. 10222712 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10222712

Brand: Thermo Scientific Acros 361832500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 261165-05-3 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261165-05-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD01320857
InChI Key RNJQBGXOSAQQDG-JGVFFNPUSA-N
Synonym 1s,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, +-1s,3r-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-n-boc-1-aminocyclopentane-3-carboxylic acid, 1s,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 1512529
IUPAC Name (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

Specifications

Melting Point 111.8°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
Quantity 250 mg
Formula Weight 229.27
Percent Purity 95%,98% ee
Physical Form Powder and/or Chunks
Chemical Name or Material (1R, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.