missing translation for 'onlineSavingsMsg'
Learn More

(1R,4S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, TRC

Product Code. 30449333
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449333 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449333 Supplier Toronto Research Chemicals Supplier No. TRCT79805025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (-)-Fenchone
CAS 7787-20-4
Molecular Formula C10 H16 O
Quantity 25 g
Synonym (-)-Fenchone, (1R,4S)-1,3,3-Trimethylnorbornan-2-one, Bicyclo[2.2.1]heptan-2-one','1,3,3-trimethyl-','(1R,4S)-, 2-Norbornanone','1,3,3-trimethyl-','(1R,4S)-(-)- (8CI), Bicyclo[2.2.1]heptan-2-one','1,3,3-trimethyl-','(1R)-, (1R)-(-)-Fenchone, (1R)-Fenchone, (1R,4S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, (R)-(-)-Fenchone, (R)-Fenchone, L-(-)-Fenchone, L-Fenchone, l-Fenchone
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m0/s1
SMILES C[C@@]12CC[C@@H](C1)C(C)(C)C2=O
IUPAC Name (1S,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
Molecular Weight (g/mol) 152.23
Formula Weight 152.1201
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.