missing translation for 'onlineSavingsMsg'
Learn More

(1R,6R)-2,8-Diazabicyclo[4.3.0]nonane (Technical Grade), TRC

Product Code. 30351485
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351485 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351485 Supplier Toronto Research Chemicals Supplier No. TRCD417275250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,6R)-2,8-Diazabicyclo[4.3.0]nonane
CAS 151213-42-2
Molecular Formula C7 H14 N2
Quantity 250 mg
Synonym 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aR,7aR)-, 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aR-cis)-, (1R,6R)-2,8-Diazabicyclo[4.3.0]nonane, (4AR,7aR)-octahydro-1H-pyrrolo[3,4-b]pyridine, (4aS,7aS)-Octahydro-6H-Pyrrolo[3,4-b]pyridine, [(4aR,7aR)-Octahydro-6H-pyrrolo[3,4-b]pyridine
InChI Formula InChI=1S/C7H14N2/c1-2-6-4-8-5-7(6)9-3-1/h6-9H,1-5H2/t6-,7+/m1/s1
SMILES C1CN[C@H]2CNC[C@H]2C1
IUPAC Name (4aR,7aR)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine
Molecular Weight (g/mol) 126.2
Formula Weight 126.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.