missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%

Product Code. 11451097
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451097 Supplier Alfa Aesar™ Supplier No. B24092.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10334-26-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863
InChI Key VNQXSTWCDUXYEZ-LDWIPMOCSA-N
Synonym 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
PubChem CID 16211789
IUPAC Name (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O

Specifications

Melting Point 197°C to 202°C
Quantity 1 g
Beilstein 2327696
Optical Rotation −102° (c=2 in Ethanol)
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Camphorquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.