missing translation for 'onlineSavingsMsg'
Learn More

(1R)-endo-(+)-Fenchyl alcohol, 96%

Product Code. 11302066
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302066 500 g 500g
11392056 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302066 Supplier Thermo Scientific Alfa Aesar Supplier No. L03211.36
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2217-02-9 | C4H4N2O5 | 160.085 g/mol

(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2217-02-9
MDL Number MFCD00003760

Specifications

Melting Point 37°C to 44°C
Boiling Point 201°C to 202°C
Quantity 500 g
Flash Point 73°C (163°F)
Odor Camphor-like
Optical Rotation +10.5° (c = 10 in Ethanol)
Molecular Formula C4H4N2O5
Beilstein 1850917
Synonym (+)-Fenchol; 1,3,3-Trimethyl-2-norbornanol
Solubility Information Insoluble in water.
InChI Key IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES CC1(C2CCC(C2)(C1O)C)C
IUPAC Name (1S,3R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-ol
Molecular Weight (g/mol) 154.25
PubChem CID 6997371
Formula Weight 154.25
Percent Purity 96%
Chemical Name or Material (1R)-endo-(+)-Fenchyl alcohol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.