missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Menthyl acetate, 98%

Product Code. 11338965
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11338965

Brand: Thermo Scientific Alfa Aesar L14095.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2623-23-6 | C12H22O2 | 198.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-23-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00001482
InChI Key XHXUANMFYXWVNG-OIKLOGQESA-N
Synonym 1r---menthyl acetate, 5r-2-isopropyl-5-methylcyclohexyl acetate, --menthylacetate, --1r-menthyl acetate
PubChem CID 12732529
IUPAC Name [(5R)-5-methyl-2-propan-2-ylcyclohexyl] acetate
SMILES CC1CCC(C(C1)OC(=O)C)C(C)C

Specifications

Density 0.925
Boiling Point 229°C to 231°C
Flash Point 77°C (170°F)
Refractive Index 1.447
Quantity 1 g
Beilstein 2208505
Merck Index 14,5839
Optical Rotation −80.5° (c=8 in Benzene)
Formula Weight 198.31
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Menthyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.