missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(+)-10-Camphorsulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (1S)-(+)-10-Camphorsulfonic Acid |
| CAS | 3144-16-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H16 O4 S |
| Quantity | 100 g |
| Synonym | Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1S,4R)-, 10-Bornanesulfonic acid, 2-oxo-, (1S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1S)-, Camphorsulfonic acid (6CI), (+)-(S)-Camphor-10-sulfonic acid, (+)-2-Oxo-10-bornanesulfonic acid, (+)-Camphor-10-sulfonic acid, (+)-Camphor-ω-sulfonic acid, (+)-Camphorsulfonic acid, (+)-D-Camphor-10-sulfonic acid, (+)-β-Camphorsulfonic acid, (1S)-(+)-10-Camphorsulfonic acid, (1S)-10-Camphorsulfonic acid, (S)-(+)-CSA, (S)-(+)-Camphor-10-sulfonic acid, (S)-Camphor-10-sulfonic acid, 2-Oxo-10-bornanesulfonic acid, Camphor-10-sulfonic acid, (+)-, Camphor-ω-sulfonic acid, (+)-, D-(+)-Camphor-10-sulfonic acid, D-(+)-Camphorsulfonic acid, D-Camphor-10-sulfonic acid, D-Camphor-β-sulfonic acid, D-Camphorsulfonic acid, Reychler's acid, d-10-Camphorsulfonic acid, d-Camphorsulfonic acid, β-Camphorsulfonic acid, (+)- |
| InChI Formula | InChI=1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m1/s1 |
| SMILES | CC1(C)[C@@H]2CC[C@@]1(CS(=O)(=O)O)C(=O)C2 |
| IUPAC Name | [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
| Molecular Weight (g/mol) | 232.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?