missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, 98% ee

Codice prodotto. 11905281 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging missing translation for 'unitSize'
11905281 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 11905281 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 362530010
 Restricted Item
Estimated Shipment: 03-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 220497-88-1 | C6H13NO3·HCl | 183.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifica

CAS 220497-88-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 124.0°C to 124.6°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H13NO3·HCl
Quantity 1 g
Synonym 1s,2r,3s,5s-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1s,2r,3s,4s-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclope, 2s,3s,5s,1r-5-amino-3-hydroxymethyl cyclopentane-1,2-diol, chloride
InChI Key BLTXEPQZAMUGID-VKYWDCQCSA-N
SMILES C1C(C(C(C1N)O)O)CO.Cl
IUPAC Name (1S,2R,3S,5S)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
Molecular Weight (g/mol) 183.63
PubChem CID 24211999
Formula Weight 183.63
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, (98% e.e.)
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • (1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.