missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, 98% ee

Product Code. 10447782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10447782 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10447782 Supplier Thermo Scientific Acros Supplier No. 362200010
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 178032-63-8 | C9H13NO3 | 183.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 178032-63-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point 150°C to 152°C
Color White
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C9H13NO3
MDL Number MFCD01320865
Quantity 1 g
Synonym 1s,2r,6s,7r-4,4-dimethyl-3,5-dioxa-8-azatricyclo 5.2.1.0 2,6 decan-9-one, 3as,4r,7s,7ar-2,2-dimethyltetrahydro-4,7-methano 1,3 dioxolo 4,5-c pyridin-6 3ah-one, 2s,7s,1r,6r-4,4-dimethyl-3,5-dioxa-9-azatricyclo 5.2.1.0<2,6> decan-8-one, 1s,2r,6s,7r-4,4-dimethyl-3,5-dioxa-8-azatricyclo 5.2.1.0 2 ,? decan-9-one, 4,7-methano-1,3-dioxolo 4,5-c pyridin-6 3ah-one,tetrahydro-2,2-dimethyl-, 3as,4r,7s,7ar
InChI Key CSGIROCFQXHTKS-WNJXEPBRSA-N
SMILES CC1(OC2C3CC(C2O1)NC3=O)C
Molecular Weight (g/mol) 183.2
PubChem CID 10219706
Formula Weight 183.2
Percent Purity 95%,98% e.e
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material (1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.