missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%

Product Code. 10598761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10598761 5 g Glass bottle 5g
10120611 25 g Glass bottle 25g
10320944 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10598761 Supplier Thermo Scientific Acros Supplier No. 334800050
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2964-48-9 | C9H12N2O4 | 212.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2964-48-9
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.2
MDL Number MFCD00007359
InChI Key OCYJXSUPZMNXEN-IUCAKERBSA-N
PubChem CID 224171
SMILES C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 160°C to 166°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5 g
Specific Rotation + 31
Synonym 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol, 1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol, 1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol, 2-amino-1-4-nitro-phenyl-propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s, threomine, 1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol, 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned, 1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s, l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol
Formula Weight 212.2
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.