missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol, 97%, Thermo Scientific Chemicals

Product Code. 10798651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10798651 5 g 5g
10206151 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10798651 Supplier Thermo Scientific Acros Supplier No. 181490050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28143-91-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004503
InChI Key JUCGVCVPNPBJIG-IUCAKERBSA-N
Synonym 1s,2s-+-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenylpropane-1,3-diol, l-+-threo-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-propane-1,3-diol, 1s, 2s-+-2-amino-1-phenyl-1,3-propanediol, 1,3-propanediol, 2-amino-1-phenyl-, 1s,2s, pubchem6415, s r*,r*-2-amino-1-phenylpropane-1,3-diol, s,s-2-amino-1-phenyl-1,3-propanediol
PubChem CID 119881
IUPAC Name (1S,2S)-2-amino-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(C(CO)N)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH(OH)CH(NH2)CH2OH
Quantity 5 g
Specific Rotation + 35.00
Formula Weight 167.21
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (1S, 2S)-(+)-2-Amino-1-phenyl-1, 3-propanediol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.