missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11975931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975931 250 mg Glass bottle 250mg
11985931 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975931 Supplier Thermo Scientific Acros Supplier No. 434092500
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1091606-67-5
Molecular Formula C26H24NP
Molecular Weight (g/mol) 381.46
MDL Number MFCD11045444
InChI Key MIPGTOGPLXZBQX-QYRYXDJWNA-N
Synonym 1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
PubChem CID 46177587
IUPAC Name (1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
SMILES N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 87.0°C to 93.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 381.45
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.