missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Methoxycyclohexanol, 97%

Product Code. 15400857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15400857 1 g 1g
11369605 5 g 5g
11379605 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 15400857

Brand: Thermo Scientific Alfa Aesar L19065.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 134108-92-2 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134108-92-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00211253
InChI Key DCQQZLGQRIVCNH-BQBZGAKWSA-N
Synonym 1s,2s-2-methoxycyclohexanol, 1s,2s-+-2-methoxycyclohexanol, cyclohexanol, 2-methoxy-, 1s,2s, 1alpha-2beta-methoxycyclohexanol, 1s,2s-trans-2-methoxycyclohexanol, 1s,2s-2-methoxycyclohexan-1-ol, 1s,2s-+-2-methoxy cyclohexanol
PubChem CID 10197706
IUPAC Name (1S,2S)-2-methoxycyclohexan-1-ol
SMILES COC1CCCCC1O

Specifications

Density 1.015
Boiling Point 195°C
Refractive Index 1.4595
Quantity 1 g
Beilstein 1848990
Optical Rotation +56° (Neat)
Formula Weight 130.19
Percent Purity 99%
Assay ee 98%
Chemical Name or Material (1S,2S)-(+)-2-Methoxycyclohexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.