missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S,3R,5S)-(+)-2,3-Pinanediol, 98%

Product Code. 10155210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155210 2.5 g Glass bottle 2.5g
10023573 10 g Glass bottle 10g
10033673 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10155210 Supplier Thermo Scientific Acros Supplier No. 279550025
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18680-27-8 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18680-27-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00077851
InChI Key MOILFCKRQFQVFS-OORONAJNSA-N
Synonym 1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s
PubChem CID 10219606
IUPAC Name (1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SMILES CC1(C2CC1C(C(C2)O)(C)O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 52°C to 60°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 06,4145
Specific Rotation Condition +8.50 (20.00°C c=6.5,toluene)
Specific Rotation + 8.5
Formula Weight 170.25
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material (1S,2S,3R,5S)-(+)-2,3-Pinanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.