missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%

Product Code. 15468886
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468886 25 g 25g
15448886 1 g 1g
15458886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468886 Supplier Thermo Scientific Alfa Aesar Supplier No. H64396.14
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 87583-89-9 | C8H18N2 | 142.246 g/mol

It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

Solubility
Slightly soluble in water.

Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87583-89-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00671528
InChI Key JRHPOFJADXHYBR-YUMQZZPRSA-N
Synonym 1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
PubChem CID 13822957
IUPAC Name (1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CNC1CCCCC1NC

Specifications

Melting Point 39°C to 44°C
Color Brown
Density 0.902 g/mL
Flash Point 74°C (165°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3259
Beilstein 4304824
Solubility Information Slightly soluble in water.
Formula Weight 142.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.