missing translation for 'onlineSavingsMsg'
Learn More

(1S,3’S)-Solifenacin Succinate (>90%), TRC

Product Code. 30409790
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409790 10 mg
30421578 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30409790 Supplier Toronto Research Chemicals Supplier No. TRCS67673010MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862207-71-4
Molecular Formula C23 H26 N2 O2 . C4 H6 O4
Molecular Weight (g/mol) 480.553
Synonym Butanedioic acid, compd. with (3S)-1-azabicyclo[2.2.2]oct-3-yl (1S)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate
IUPAC Name [(3S)-1-azabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;butanedioic acid
SMILES OC(=O)CCC(=O)O.O=C(O[C@@H]1C[N@@]2CC[C@H]1CC2)N3CCc4ccccc4[C@@H]3c5ccccc5

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H26N2O2.C4H6O4/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;5-3(6)1-2-4(7)8/h1-9,18,21-22H,10-16H2;1-2H2,(H,5,6)(H,7,8)/t21-,22+;/m1./s1
Formula Weight 480.226
Chemical Name or Material (1S,3’S)-Solifenacin Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.