missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid, 95%

Product Code. 15459535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15459535 250 mg 250mg
15449535 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15459535

Brand: Thermo Scientific Alfa Aesar H52040.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71830-07-4 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71830-07-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00211290
InChI Key MLLSSTJTARJLHK-CRCLSJGQSA-N
Synonym 1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-aminocyclopentane-1-carboxylic acid, +-1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-amino-cyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, 1s,3r-+-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1s,3r, pubchem18730, +-cacp, cis-3-aminocyclopentane-1-carboxylic acid
PubChem CID 1502034
IUPAC Name (1S,3R)-3-aminocyclopentane-1-carboxylic acid
SMILES C1CC(CC1C(=O)O)N

Specifications

Quantity 250 mg
Formula Weight 129.16
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.