missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-ACPD, R&D Systems™

Product Code. 30001071
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001071 10 mg 10mg
30001072 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001071 Supplier Tocris Bioscience Supplier No. 0284/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Group I and group II mGlu agonist; active isomer of (±)-trans-ACPD (Cat. No. 0187)

Active isomer of (±)-trans-ACPD . Agonist at both group I and II mGlu receptors (EC50 values are 5, 15, 42 and 60 μM at mGluR2, mGluR5, mGluR1 and mGluR6 respectively).

NPEC-caged-(1S,3R)-ACPD, (±)-trans-ACPD and cis-ACPD also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111900-32-4
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
InChI Key YFYNOWXBIBKGHB-FBCQKBJTSA-N
Synonym 1s,3r-1-aminocyclopentane-1,3-dicarboxylic acid, 1s,3r-acpd, trans-acpd, 1-amino-1,3-dicarboxycyclopentane, trans-+/--acpd, chembl34453, 1s,3s-acpd, 1-aminocyclopentane-1s,3r-dicarboxylic acid, 1s,3r-1-acpd
PubChem CID 104766
IUPAC Name (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
SMILES C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 173.17
Percent Purity >98%
Product Line Tocris
Chemical Name or Material (1S, 3R)-ACPD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.