missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-(-)-Camphoric acid, 98%

Product Code. 15490468
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490468 5 g 5g
15400478 250 mg 250mg
15480468 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490468 Supplier Thermo Scientific Alfa Aesar Supplier No. H54042.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 560-09-8 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 560-09-8
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00064937
InChI Key LSPHULWDVZXLIL-QUBYGPBYSA-N
Synonym 1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid
PubChem CID 219463
IUPAC Name (1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
SMILES CC1(C(CCC1(C)C(=O)O)C(=O)O)C

Specifications

Melting Point 188°C to 190°C
Quantity 5 g
Optical Rotation -48° (c=10 in Ethanol)
Beilstein 1874674
Merck Index 141733
Formula Weight 200.23
Percent Purity 98%
Chemical Name or Material (1S,3R)-(-)-Camphoric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.