missing translation for 'onlineSavingsMsg'
Learn More

(1S)-4,5-Dimethoxy-1-[(methylamino)methyl]benzocyclobutane Hydrochloride, TRC

Product Code. 30388644
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388644 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388644 Supplier Toronto Research Chemicals Supplier No. TRCD461760250MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S)-4,5-Dimethoxy-1-[(methylamino)methyl]benzocyclobutane hydrochloride
CAS 866783-13-3
Molecular Formula C12 H17 N O2 . Cl H
Quantity 250 mg
Synonym Bicyclo[4.2.0]octa-1,3,5-triene-7-methanamine, 3,4-dimethoxy-N-methyl-, hydrochloride (1:1), (7S)-, Bicyclo[4.2.0]octa-1,3,5-triene-7-methanamine, 3,4-dimethoxy-N-methyl-, hydrochloride, (7S)- (9CI), (1S)-4,5-Dimethoxy-1-[(methylamino)methyl]benzocyclobutane hydrochloride, 1-[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]-N-methylmethanamine Hydrochloride
InChI Formula InChI=1S/C12H17NO2.ClH/c1-13-7-9-4-8-5-11(14-2)12(15-3)6-10(8)9;/h5-6,9,13H,4,7H2,1-3H3;1H/t9-;/m1./s1
SMILES Cl.CNC[C@H]1Cc2cc(OC)c(OC)cc12
IUPAC Name 1-[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]-N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 243.73
Formula Weight 243.1026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.