missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, Thermo Scientific™

Product Code. 10109991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10109991 250 mg Glass bottle 250mg
10498162 1 g Glass bottle 1g
11966621 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10109991 Supplier Thermo Scientific Acros Supplier No. 362222500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 134003-03-5
InChI Key UIVLZOWDXYXITH-UHNVWZDZSA-N
Synonym 1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
PubChem CID 2734523
IUPAC Name (1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
SMILES C1CC2CC1C(=O)N2

Specifications

Melting Point 76.2°C
Color White to Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H9NO
Quantity 250 mg
Molecular Weight (g/mol) 111.14
Formula Weight 111.14
Percent Purity 95%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one
Show More Show Less

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.