missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%

Product Code. 10693975 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10693975 1 g Glass bottle 1g
10597054 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10693975 Supplier Thermo Scientific Acros Supplier No. 398040010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 113451-59-5 | C10H18N2O2 | 198.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113451-59-5
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.26
MDL Number MFCD01569250
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
PubChem CID 11521263
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 74.0°C to 76.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -41° to -47° (20°C, 589nm) (C=1, chloroform)
Solubility Information Solubility in water: insoluble
Formula Weight 198.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane
Show More Show Less
compliance-icons
Product Identifier
  • (1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.