missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%

Product Code. 11394576
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394576 1 g 1g
11314586 250 mg 250mg
11304586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394576 Supplier Thermo Scientific Alfa Aesar Supplier No. L20131.03
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 113451-59-5 | C10H18N2O2 | 198.266 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113451-59-5
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.266
MDL Number MFCD01569250
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
PubChem CID 11521263
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Beilstein 6460311
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation −44° (c=1 in Chloroform)
Formula Weight 198.26
Percent Purity 97%
Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.